Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: anxa1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181758.1 | NP_861423.1 | 2 - 3 | 126 | 71 | |
XM_009301449.1 | XP_009299724.1 | 2 - 3 | 182 | 71 | ||
GenBank mRNA | AY178793.1 | - | 2 - 3 | 126 | 71 | |
BC053190.1 | - | 1 - 2 | 109 | 71 | ||
BC164376.1 | - | 1 - 2 | 107 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |