Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: anxa1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181758.1 | NP_861423.1 | 6 - 7 | 504 | 121 | |
XM_009301449.1 | XP_009299724.1 | 6 - 7 | 560 | 121 | ||
GenBank mRNA | AY178793.1 | - | 6 - 7 | 504 | 121 | |
BC053190.1 | - | 5 - 6 | 487 | 121 | ||
BC164376.1 | - | 5 - 6 | 485 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |