Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194381.2 | NP_919362.2 | 3 - 4 | 852 | 90 | |
GenBank mRNA | AY283178.1 | - | 3 - 4 | 879 | 90 | |
BC092681.1 | - | 3 - 4 | 852 | 90 | ||
BC152122.1 | - | 3 - 4 | 851 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |