Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppp2r5ea | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194415.2 | NP_919396.1 | 12 - 13 | 1657 | 97 | |
GenBank mRNA | AY150214.1 | - | 11 - 12 | 1204 | 97 | |
BC044398.1 | - | 12 - 13 | 1473 | 97 | ||
BC078645.1 | - | 12 - 13 | 1657 | 97 | ||
BC165347.1 | - | 11 - 12 | 1238 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |