Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppp2r5ea | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194415.2 | NP_919396.1 | 5 - 6 | 997 | 75 | |
GenBank mRNA | AY150214.1 | - | 4 - 5 | 544 | 75 | |
BC044398.1 | - | 5 - 6 | 813 | 75 | ||
BC078645.1 | - | 5 - 6 | 997 | 75 | ||
BC165347.1 | - | 4 - 5 | 578 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |