Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: p2rx3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198986.3 | NP_945337.3 | 8 - 9 | 1034 | 83 | |
XM_021466489.1 | XP_021322164.1 | 8 - 9 | 1100 | 83 | ||
GenBank mRNA | AY292651.1 | - | 8 - 9 | 863 | 83 | |
BC095293.1 | - | 8 - 9 | 1035 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |