Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: evla | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199519.3 | NP_955813.3 | 6 - 7 | 1128 | 91 | |
XM_005158792.4 | XP_005158849.1 | 7 - 8 | 1754 | 91 | ||
XM_005158793.4 | XP_005158850.1 | 7 - 8 | 1747 | 91 | ||
XM_005158794.4 | XP_005158851.1 | 7 - 8 | 1542 | 91 | ||
XM_005158795.4 | XP_005158852.1 | 7 - 8 | 1881 | 91 | ||
XM_005158796.4 | XP_005158853.1 | 5 - 6 | 1458 | 91 | ||
XM_005158797.4 | XP_005158854.1 | 6 - 7 | 995 | 91 | ||
XM_005158798.4 | XP_005158855.1 | 6 - 7 | 993 | 91 | ||
XM_005158799.4 | XP_005158856.1 | 6 - 7 | 990 | 91 | ||
XM_021467223.1 | XP_021322898.1 | 7 - 8 | 1752 | 91 | ||
GenBank mRNA | BC056325.1 | - | 6 - 7 | 1099 | 91 | |
BC065353.1 | - | 6 - 7 | 1092 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |