Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: setd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199600.1 | NP_955894.1 | 8 - 9 | 1215 | 120 | |
GenBank mRNA | BC044440.1 | - | 8 - 9 | 1231 | 120 | |
DQ851816.1 | - | 1 - 2 | 145 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |