Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cul3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199691.2 | NP_955985.2 | 4 - 5 | 692 | 121 | |
GenBank mRNA | AY423034.1 | - | 4 - 5 | 685 | 121 | |
BC065357.1 | - | 4 - 5 | 654 | 121 | ||
EH455670.1 | - | 4 - 5 | 688 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |