Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: esf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199745.2 | NP_956039.2 | 5 - 6 | 1013 | 62 | |
GenBank mRNA | BC054666.1 | - | 649 | 62 | ||
BC080208.1 | - | 5 - 6 | 983 | 62 | ||
BC153482.1 | - | 3 - 4 | 637 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |