Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: larp6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199903.1 | NP_956197.1 | 1 - 2 | 372 | 112 | |
GenBank mRNA | BC056595.1 | - | 1 - 2 | 372 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |