Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199942.2 | NP_956236.1 | 2 - 3 | 488 | 146 | |
GenBank mRNA | BC057430.1 | - | 398 | 146 | ||
EH585004.1 | - | 2 - 3 | 446 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |