Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lmbrd2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199944.1 | NP_956238.1 | 5 - 6 | 637 | 78 | |
GenBank mRNA | BC063324.1 | - | 5 - 6 | 637 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |