Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vil1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200238.1 | NP_956532.1 | 7 - 8 | 851 | 94 | |
XM_005167920.4 | XP_005167977.1 | 7 - 8 | 929 | 94 | ||
GenBank mRNA | BC047186.1 | - | 7 - 8 | 851 | 94 | |
BC165692.1 | - | 6 - 7 | 804 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |