Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rasl12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200395.1 | NP_956689.1 | 2 - 3 | 281 | 82 | |
XM_017356827.2 | XP_017212316.1 | 2 - 3 | 475 | 82 | ||
GenBank mRNA | BC054128.1 | - | 2 - 3 | 281 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |