Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hif1al | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200405.1 | NP_956699.1 | 10 - 11 | 1413 | 57 | |
XM_005157571.4 | XP_005157628.2 | 10 - 11 | 57 | |||
XM_005157572.4 | XP_005157629.2 | 10 - 11 | 57 | |||
XM_021481358.1 | XP_021337033.1 | 8 - 9 | 57 | |||
GenBank mRNA | AY330295.1 | - | 10 - 11 | 1441 | 57 | |
BC054582.1 | - | 10 - 11 | 1413 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |