Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hsd17b12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200881.1 | NP_957175.1 | 5 - 6 | 597 | 69 | |
GenBank mRNA | AY551082.1 | - | 5 - 6 | 590 | 69 | |
BC048053.1 | - | 5 - 6 | 539 | 69 | ||
BC063943.1 | - | 5 - 6 | 597 | 69 | ||
BC164345.1 | - | 5 - 6 | 514 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |