Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ubtfl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201003.1 | NP_957297.1 | 2 - 3 | 160 | 63 | |
XM_005163486.3 | XP_005163543.1 | 2 - 3 | 1085 | 63 | ||
XM_005163487.3 | XP_005163544.1 | 2 - 3 | 1085 | 63 | ||
XM_005163488.3 | XP_005163545.1 | 2 - 3 | 1085 | 63 | ||
XM_005163489.3 | XP_005163546.1 | 2 - 3 | 1085 | 63 | ||
XM_005163490.3 | XP_005163547.1 | 2 - 3 | 1085 | 63 | ||
XM_009298612.2 | XP_009296887.1 | 2 - 3 | 1085 | 63 | ||
XM_009298613.2 | XP_009296888.1 | 2 - 3 | 1085 | 63 | ||
XM_009298614.2 | XP_009296889.1 | 2 - 3 | 1085 | 63 | ||
XM_017357972.2 | XP_017213461.1 | 1 - 2 | 111 | 63 | ||
GenBank mRNA | BC055140.1 | - | 2 - 3 | 160 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |