Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rps3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201153.1 | NP_957447.1 | 1 - 2 | 63 | 68 | |
GenBank mRNA | AY394937.1 | - | 1 - 2 | 37 | 68 | |
BC045902.1 | - | 1 - 2 | 63 | 68 | ||
BC165099.1 | - | 1 - 2 | 70 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |