Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dcun1d2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205538.1 | NP_991101.1 | 2 - 3 | 237 | 81 | |
XM_005167810.4 | XP_005167867.1 | 2 - 3 | 342 | 81 | ||
XM_005167811.4 | XP_005167868.1 | 2 - 3 | 364 | 81 | ||
XM_021478598.1 | XP_021334273.1 | 2 - 3 | 309 | 81 | ||
GenBank mRNA | BC065884.1 | - | 2 - 3 | 237 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |