Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rlbp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205690.2 | NP_991253.1 | 3 - 4 | 379 | 134 | |
XM_005163003.4 | XP_005163060.1 | 4 - 5 | 462 | 134 | ||
GenBank mRNA | BC065863.1 | - | 3 - 4 | 383 | 134 | |
EU348854.1 | - | 2 - 3 | 349 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |