Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ctsla | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212584.1 | NP_997749.1 | 4 - 5 | 638 | 78 | |
XM_005165268.2 | XP_005165325.1 | 5 - 6 | 776 | 78 | ||
XM_021476007.1 | XP_021331682.1 | 5 - 6 | 985 | 78 | ||
GenBank mRNA | AF498292.1 | - | 3 - 4 | 548 | 78 | |
BC066490.1 | - | 4 - 5 | 638 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Probe spans exons |