Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213002.1 | NP_998167.1 | 7 - 8 | 878 | 104 | |
GenBank mRNA | AY648785.1 | - | 7 - 8 | 892 | 104 | |
BC051777.1 | - | 7 - 8 | 878 | 104 | ||
BC065610.1 | - | 7 - 8 | 862 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |