Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213002.1 | NP_998167.1 | 8 - 9 | 1026 | 88 | |
GenBank mRNA | BC051777.1 | - | 8 - 9 | 1026 | 88 | |
BC065610.1 | - | 8 - 9 | 1010 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |