Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mospd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213369.1 | NP_998534.1 | 5 - 6 | 784 | 125 | |
GenBank mRNA | BC046085.1 | - | 5 - 6 | 784 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |