Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stmn4l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002563.1 | NP_001002563.1 | 3 - 4 | 215 | 110 | |
XM_005158637.4 | XP_005158694.1 | 3 - 4 | 236 | 110 | ||
GenBank mRNA | BC076237.1 | - | 3 - 4 | 215 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |