Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stmn4l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002563.1 | NP_001002563.1 | 4 - 5 | 436 | 131 | |
XM_005158637.4 | XP_005158694.1 | 4 - 5 | 457 | 131 | ||
GenBank mRNA | BC076237.1 | - | 4 - 5 | 436 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |