Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aarsd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003572.1 | NP_001003572.1 | 2 - 3 | 238 | 132 | |
GenBank mRNA | BC077118.1 | - | 2 - 3 | 238 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |