Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aarsd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003572.1 | NP_001003572.1 | 8 - 9 | 928 | 90 | |
GenBank mRNA | BC077118.1 | - | 8 - 9 | 928 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |