Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atp6v1c1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005772.2 | NP_001005772.2 | 3 - 4 | 469 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Danio rerio Probe spans exons |