Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atp6v1c1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005772.2 | NP_001005772.2 | 5 - 6 | 639 | 135 | |
GenBank mRNA | BC083532.1 | - | 5 - 6 | 543 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |