Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpr34a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007215.1 | NP_001007216.1 | 1049 | 63 | ||
GenBank mRNA | BC162726.1 | - | 1092 | 63 | ||
BC162729.1 | - | 1092 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Mammalian Gene Collection Recommended Assay Danio rerio |