Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gnal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007339.2 | NP_001007340.1 | 9 - 10 | 980 | 154 | |
GenBank mRNA | BC085540.1 | - | 9 - 10 | 983 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |