Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rhoua | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007443.1 | NP_001007444.1 | 1 - 2 | 643 | 57 | |
GenBank mRNA | AY865564.1 | - | 1 - 2 | 245 | 57 | |
BC085398.1 | - | 1 - 2 | 643 | 57 | ||
BC165147.1 | - | 1 - 2 | 288 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |