Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rhoua | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007443.1 | NP_001007444.1 | 2 - 3 | 704 | 59 | |
GenBank mRNA | AY865564.1 | - | 2 - 3 | 306 | 59 | |
BC085398.1 | - | 2 - 3 | 704 | 59 | ||
BC165147.1 | - | 2 - 3 | 349 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |