Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mfsd2aa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007451.1 | NP_001007452.1 | 8 - 9 | 1098 | 89 | |
XM_005156993.4 | XP_005157050.1 | 9 - 10 | 1184 | 89 | ||
GenBank mRNA | BC085388.1 | - | 8 - 9 | 1098 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |