Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: usp2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008574.2 | NP_001008574.2 | 9 - 10 | 1149 | 86 | |
XM_005157513.4 | XP_005157570.1 | 11 - 12 | 1652 | 86 | ||
XM_005157514.4 | XP_005157571.1 | 11 - 12 | 1746 | 86 | ||
XM_005157515.4 | XP_005157572.1 | 11 - 12 | 1644 | 86 | ||
XM_005157516.4 | XP_005157573.1 | 9 - 10 | 1220 | 86 | ||
XM_017350951.2 | XP_017206440.1 | 11 - 12 | 1647 | 86 | ||
XM_021481396.1 | XP_021337071.1 | 11 - 12 | 1750 | 86 | ||
GenBank mRNA | BC086963.1 | - | 9 - 10 | 990 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |