Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gcsha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013457.1 | NP_001013475.1 | 1 - 2 | 190 | 86 | |
GenBank mRNA | BC091548.1 | - | 1 - 2 | 190 | 86 | |
BC164865.1 | - | 1 - 2 | 188 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |