Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bccip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013475.2 | NP_001013493.2 | 1 - 2 | 190 | 74 | |
GenBank mRNA | BC091454.1 | - | 1 - 2 | 144 | 74 | |
BC164248.1 | - | 1 - 2 | 162 | 74 | ||
EH457160.1 | - | 1 - 2 | 224 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |