Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hmgcrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014292.2 | NP_001014314.1 | 17 - 18 | 2413 | 72 | |
GenBank mRNA | BC091792.1 | - | 17 - 18 | 2417 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |