Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: syt5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020546.2 | NP_001018382.2 | 6 - 7 | 903 | 82 | |
GenBank mRNA | BC095109.1 | - | 6 - 7 | 903 | 82 | |
BC165563.1 | - | 5 - 6 | 820 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |