Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acbd5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020647.1 | NP_001018483.1 | 6 - 7 | 679 | 84 | |
XM_005162509.4 | XP_005162566.1 | 6 - 7 | 84 | |||
XM_009297238.3 | XP_009295513.1 | 7 - 8 | 84 | |||
XM_009297239.3 | XP_009295514.1 | 7 - 8 | 84 | |||
XM_009297241.3 | XP_009295516.1 | 7 - 8 | 84 | |||
XM_021470173.1 | XP_021325848.1 | 7 - 8 | 84 | |||
XM_021470174.1 | XP_021325849.1 | 7 - 8 | 84 | |||
GenBank mRNA | BC095655.1 | - | 6 - 7 | 679 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |