Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ahr1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024816.2 | NP_001019987.1 | 3 - 4 | 809 | 78 | |
GenBank mRNA | DQ016494.1 | - | 3 - 4 | 809 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Danio rerio Probe spans exons |