Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wwtr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037696.1 | NP_001032785.1 | 4 - 5 | 990 | 80 | |
XM_005161848.4 | XP_005161905.1 | 4 - 5 | 1348 | 80 | ||
GenBank mRNA | BC162350.1 | - | 4 - 5 | 917 | 80 | |
BC162832.1 | - | 4 - 5 | 917 | 80 | ||
DQ238862.1 | - | 4 - 5 | 990 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |