Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: anpepa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044861.1 | NP_001038326.1 | 2 - 3 | 732 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Danio rerio Probe spans exons |