Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cspg5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044892.1 | NP_001038357.1 | 3 - 4 | 1497 | 83 | |
XM_005159515.4 | XP_005159572.2 | 3 - 4 | 1562 | 83 | ||
XM_005159516.4 | XP_005159573.2 | 3 - 4 | 1560 | 83 | ||
XM_009294162.3 | XP_009292437.1 | 3 - 4 | 1562 | 83 | ||
GenBank mRNA | BC135055.1 | - | 2 - 3 | 655 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |