Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kctd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045333.2 | NP_001038798.2 | 2 - 3 | 605 | 79 | |
GenBank mRNA | BC118684.1 | - | 2 - 3 | 608 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |