Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045457.1 | NP_001038922.1 | 3 - 4 | 625 | 79 | |
GenBank mRNA | BC122228.1 | - | 3 - 4 | 625 | 79 | |
BC165445.1 | - | 3 - 4 | 502 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |