Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc35f2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076556.1 | NP_001070024.1 | 5 - 6 | 655 | 80 | |
GenBank mRNA | BC124339.1 | - | 5 - 6 | 655 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Inventoried Probe spans exons |