Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: col27a1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080575.1 | NP_001074044.1 | 29 - 30 | 2648 | 65 | |
XM_017356118.2 | XP_017211607.1 | 29 - 30 | 3066 | 65 | ||
XM_021476201.1 | XP_021331876.1 | 9 - 10 | 428 | 65 | ||
GenBank mRNA | EF032484.1 | - | 29 - 30 | 2648 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Danio rerio Probe spans exons |